Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145645.2 | NP_001139117.1 | 5 | 1077 | ||
NM_006573.4 | NP_006564.1 | 6 | 1134 | |||
GenBank mRNA | AF116456.1 | - | 6 | 985 | ||
AF132600.1 | - | 6 | 1003 | |||
AF134715.1 | - | 6 | 1011 | |||
AF186114.1 | - | 6 | 1024 | |||
AK309629.1 | - | 7 | 904 | |||
AY358881.1 | - | 6 | 1036 | |||
BC020674.1 | - | 6 | 1046 | 4719315 | ||
D79690.1 | - | 2 | 204 |