Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006580.3 | NP_006571.1 | 3 | 793 | ||
GenBank mRNA | AF152101.1 | - | 3 | 613 | ||
AK314955.1 | - | 3 | 792 | |||
BC069662.1 | - | 3 | 613 | 7262563 | ||
BC069682.1 | - | 3 | 613 | 7262480 | ||
BC069759.1 | - | 3 | 613 | 7262492 | ||
BC069777.1 | - | 3 | 613 | 7262504 |