Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNPY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318842.1 | NP_001305771.1 | 4 | 865 | ||
NM_001318845.1 | NP_001305774.1 | 2 | 642 | |||
NM_001318847.1 | NP_001305776.1 | 4 | 828 | |||
NM_001318848.1 | NP_001305777.1 | 3 | 766 | |||
NM_006586.4 | NP_006577.2 | 3 | 766 | |||
NR_134880.1 | 4 | 828 | ||||
NR_134881.1 | 5 | 2021 | ||||
NR_134882.1 | 3 | 704 | ||||
NR_134885.1 | 3 | 863 | ||||
NR_134886.1 | 2 | 642 | ||||
NR_134888.1 | 2 | 642 | ||||
GenBank mRNA | AK090425.1 | - | 1 | 1190 | ||
AK124196.1 | - | 5 | 2020 | |||
AY358960.1 | - | 3 | 661 | |||
BC004423.1 | - | 3 | 712 | 2819660 | ||
BC008133.1 | - | 2 | 278 | |||
BC008898.2 | - | 3 | 393 | 2959532 | ||
BC008961.2 | - | 3 | 393 | 2959532 | ||
BF528267.1 | - | 2 | 234 | |||
BG675987.1 | - | 4 | 422 | |||
BI909527.1 | - | 3 | 356 | |||
BQ063472.1 | - | 4 | 471 | |||
BQ680146.1 | - | 3 | 385 | |||
CK429647.1 | - | 4 | 441 | |||
CN302713.1 | - | 3 | 504 | |||
HY101691.1 | - | 2 | 290 |
Gene Symbol: CNPY3-GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134890.1 | 2 | 642 | |||
GenBank mRNA | FY213816.1 | - | 2 | 174 |