Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNPY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318842.1 | NP_001305771.1 | 4, 5 | 904 | ||
NM_001318845.1 | NP_001305774.1 | 2, 3 | 681 | |||
NM_006586.4 | NP_006577.2 | 3, 4 | 805 | |||
NR_134880.1 | 4, 5 | 867 | ||||
NR_134881.1 | 5, 6 | 2060 | ||||
NR_134882.1 | 3, 4 | 743 | ||||
GenBank mRNA | AK124196.1 | - | 5, 6 | 2059 | ||
AY358960.1 | - | 3, 4 | 700 | |||
BC004423.1 | - | 3, 4 | 751 | 2819660 | ||
BC008133.1 | - | 2, 3 | 317 | |||
BC008898.2 | - | 3, 4 | 432 | 2959532 | ||
BC008961.2 | - | 3, 4 | 432 | 2959532 | ||
BG675987.1 | - | 4, 5 | 461 | |||
BI909527.1 | - | 3, 4 | 395 | |||
BQ063472.1 | - | 4, 5 | 510 | |||
U80744.1 | - | 3, 4 | 750 |