Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006606.2 | NP_006597.2 | 5 | 705 | ||
XM_005260652.2 | XP_005260709.1 | 4 | 669 | |||
GenBank mRNA | AF237576.1 | - | 5 | 659 | ||
AL832411.1 | - | 5 | 653 | |||
BC015938.1 | - | 5 | 705 | 3922423 |