Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253387.1 | NP_001240316.1 | 5 | 890 | ||
NM_001253388.1 | NP_001240317.1 | 3 | 342 | |||
NM_006614.3 | NP_006605.2 | 5 | 890 | |||
XM_006712938.1 | XP_006713001.1 | 5 | 501 | |||
XM_006712939.3 | XP_006713002.1 | 4 | 818 | |||
XM_006712940.3 | XP_006713003.1 | 4 | 732 | |||
XM_011533292.1 | XP_011531594.1 | 5 | 871 | |||
XM_011533294.1 | XP_011531596.1 | 5 | 501 | |||
XM_011533295.1 | XP_011531597.1 | 5 | 871 | |||
XM_011533296.1 | XP_011531598.1 | 5 | 501 | |||
XM_017005566.1 | XP_016861055.1 | 6 | 5395 | |||
XM_017005567.1 | XP_016861056.1 | 4 | 603 | |||
XM_017005568.1 | XP_016861057.1 | 4 | 410 | |||
XM_017005569.1 | XP_016861058.1 | 4 | 818 | |||
XM_017005570.1 | XP_016861059.1 | 4 | 732 | |||
XM_017005571.1 | XP_016861060.1 | 4 | 603 | |||
XM_017005572.1 | XP_016861061.1 | 5 | 898 | |||
XM_017005573.1 | XP_016861062.1 | 4 | 818 | |||
GenBank mRNA | AB209329.1 | - | 5 | 715 | ||
AF002246.1 | - | 5 | 519 | |||
AK126878.1 | - | 5 | 869 | |||
BC104918.1 | - | 3 | 279 | 8143921 | ||
BC143496.1 | - | 3 | 276 | 9052004 |