Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319676.1 | NP_001306605.1 | 18 | 2087 | ||
NM_006615.2 | NP_006606.1 | 20 | 2276 | |||
NM_016452.2 | NP_057536.1 | 19 | 2198 | |||
XM_011544017.1 | XP_011542319.1 | 19 | 2201 | |||
XM_011544018.1 | XP_011542320.1 | 18 | 2123 | |||
XM_011544019.2 | XP_011542321.1 | 17 | 2123 | |||
XM_011544020.1 | XP_011542322.1 | 19 | 1947 | |||
XM_017000098.1 | XP_016855587.1 | 18 | 2117 | |||
XM_017000099.1 | XP_016855588.1 | 20 | 2006 | |||
GenBank mRNA | AB038463.1 | - | 19 | 2167 | ||
AF022799.1 | - | 20 | 2276 | |||
BC027993.1 | - | 18 | 2038 | 5173512 |