Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003677.4 | NP_003668.2 | 7 | 630 | ||
GenBank mRNA | AB014731.1 | - | 7 | 601 | ||
AF038554.1 | - | 7 | 626 | |||
AF103800.1 | - | 4 | 349 | |||
BC007860.2 | - | 7 | 560 | 4300744 |