Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285460.1 | NP_001272389.1 | 7 | 603 | ||
NM_006633.3 | NP_006624.2 | 8 | 997 | |||
XM_005248410.2 | XP_005248467.1 | 8 | 847 | |||
XM_017008960.1 | XP_016864449.1 | 8 | 946 | |||
GenBank mRNA | AK291066.1 | - | 8 | 997 | ||
AK302510.1 | - | 7 | 603 | |||
AK307555.1 | - | 8 | 771 | |||
U51903.1 | - | 8 | 928 |