Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WASF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291965.1 | NP_001278894.1 | 5 | 611 | ||
NM_006646.5 | NP_006637.2 | 5 | 611 | |||
XM_011534889.1 | XP_011533191.1 | 5 | 622 | |||
XM_011534890.1 | XP_011533192.1 | 4 | 552 | |||
GenBank mRNA | AB020707.2 | - | 4 | 500 | ||
AB026543.1 | - | 3 | 386 | |||
AF134305.1 | - | 2 | 245 | |||
AF454702.1 | - | 5 | 564 | |||
AK289845.1 | - | 5 | 611 | |||
BC050283.1 | - | 5 | 611 | 5300292 |