Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WASF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291965.1 | NP_001278894.1 | 9 | 1218 | ||
NM_006646.5 | NP_006637.2 | 9 | 1227 | |||
XM_011534889.1 | XP_011533191.1 | 9 | 1238 | |||
XM_011534890.1 | XP_011533192.1 | 8 | 1168 | |||
GenBank mRNA | AB020707.2 | - | 8 | 1116 | ||
AB026543.1 | - | 7 | 1002 | |||
AF134305.1 | - | 6 | 861 | |||
AF454702.1 | - | 9 | 1180 | |||
AK289845.1 | - | 9 | 1227 | |||
AK294725.1 | - | 3 | 339 | |||
BC050283.1 | - | 9 | 1218 | 5300292 |