Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEU3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006656.5 | NP_006647.3 | 2 | 451 | ||
GenBank mRNA | AB008185.2 | - | 3 | 366 | ||
AK022450.1 | - | 2 | 445 | |||
AK290442.1 | - | 2 | 413 | |||
BC136397.1 | - | 3 | 785 | 9020384 | ||
BC144059.1 | - | 3 | 785 | 9052579 | ||
CN429425.1 | - | 2 | 361 | |||
DB240271.1 | - | 2 | 451 | |||
Y18563.1 | - | 2 | 1222 |