Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145123.2 | NP_001138595.1 | 4 | 781 | ||
NM_006658.4 | NP_006649.2 | 5 | 934 | |||
GenBank mRNA | AF071789.1 | - | 4 | 659 | ||
AF097730.1 | - | 4 | 656 | |||
AK293478.1 | - | 4 | 583 | |||
BC028094.1 | - | 5 | 718 | 5241722 |