Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003683.5 | NP_003674.1 | 2 | 263 | ||
XM_017028485.1 | XP_016883974.1 | 2 | 254 | |||
XM_017028486.1 | XP_016883975.1 | 2 | 249 | |||
GenBank mRNA | AK223185.1 | - | 2 | 224 | ||
AK223215.1 | - | 2 | 220 | |||
AK303000.1 | - | 2 | 223 | |||
AK313116.1 | - | 2 | 220 | |||
BC000380.2 | - | 2 | 210 | 2820455 | ||
BC014787.1 | - | 2 | 185 | |||
BG829556.1 | - | 251 | ||||
DA275615.1 | - | 2 | 265 | |||
U79775.1 | - | 2 | 208 |