Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006660.4 | NP_006651.2 | 2 | 459 | ||
NR_133680.1 | 2 | 459 | ||||
XM_011521164.2 | XP_011519466.1 | 2 | 443 | |||
GenBank mRNA | AJ006267.3 | - | 2 | 264 | ||
AK292316.1 | - | 2 | 385 | |||
AK308133.1 | - | 2 | 379 | |||
AK310614.1 | - | 2 | 381 | |||
AL136922.1 | - | 2 | 362 | |||
BC130373.1 | - | 2 | 255 | 40146404 | ||
BC136487.1 | - | 2 | 255 | 9020476 | ||
BG717994.1 | - | 2 | 464 |