Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008563.4 | NP_001008563.2 | 6 | 550 | ||
NM_001110303.3 | NP_001103773.2 | 6 | 452 | |||
NM_006676.7 | NP_006667.3 | 6 | 452 | |||
XM_005251665.3 | XP_005251722.1 | 6 | 440 | |||
XM_005251666.3 | XP_005251723.1 | 2 | 93 | |||
XM_011518161.2 | XP_011516463.1 | 7 | 792 | |||
XM_011518162.2 | XP_011516464.1 | 6 | 971 | |||
XM_017014221.1 | XP_016869710.1 | 2 | 93 | |||
GenBank mRNA | AB023220.1 | - | 5 | 373 | ||
AY074877.1 | - | 6 | 489 | |||
BC039593.1 | - | 6 | 399 | 5588842 | ||
BF727180.1 | - | 6 | 465 | |||
BP246486.1 | - | 6 | 440 | |||
DA791069.1 | - | 6 | 440 |