Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131027.1 | NP_001124499.1 | 6 | 945 | ||
NM_003687.3 | NP_003678.2 | 7 | 1063 | |||
XM_017010002.1 | XP_016865491.1 | 5 | 816 | |||
GenBank mRNA | AF153882.1 | - | 7 | 1027 | ||
AF154335.1 | - | 7 | 1050 | |||
AK225407.1 | - | 6 | 1349 | |||
BC003096.1 | - | 6 | 896 | 3501794 | ||
BC016765.1 | - | 7 | 1065 | 4132234 | ||
X93510.1 | - | 7 | 1021 |