Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006688.4 | NP_006679.1 | 1 | 698 | ||
GenBank mRNA | AF095154.1 | - | 1 | 483 | ||
BC008798.2 | - | 1 | 698 | 3635430 |