Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006699.3 | NP_006690.1 | 13 | 4345 | ||
XM_006710302.2 | XP_006710365.1 | 14 | 4041 | |||
GenBank mRNA | AK090840.1 | - | 1 | 1213 | ||
BC063300.1 | - | 13 | 4343 | 5263292 |