Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003693.3 | NP_003684.2 | 2 | 244 | ||
NM_145350.2 | NP_663325.1 | 2 | 244 | |||
NR_028075.2 | 2 | 244 | ||||
NR_102409.1 | 2 | 281 | ||||
GenBank mRNA | AB052947.1 | - | 2 | 178 | ||
AB052948.1 | - | 2 | 211 | |||
AB052949.1 | - | 2 | 176 | |||
AB052950.1 | - | 2 | 168 | |||
AK291810.1 | - | 2 | 187 | |||
AK310174.1 | - | 2 | 204 | |||
BC039735.1 | - | 2 | 212 | 6041360 | ||
D63483.1 | - | 2 | 154 | |||
D86864.1 | - | 2 | 178 |