Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286625.1 | NP_001273554.1 | 8 | 2032 | ||
NM_001286626.1 | NP_001273555.1 | 7 | 2039 | |||
NM_001286627.1 | NP_001273556.1 | 6 | 1897 | |||
NM_006711.4 | NP_006702.1 | 8 | 2093 | |||
NM_080594.3 | NP_542161.1 | 8 | 2227 | |||
NR_104485.1 | 7 | 2161 | ||||
XM_005255048.1 | XP_005255105.1 | 8 | 2320 | |||
XM_005255049.3 | XP_005255106.1 | 8 | 3785 | |||
XM_017022874.1 | XP_016878363.1 | 8 | 2686 | |||
GenBank mRNA | AF015608.1 | - | 7 | 1777 | ||
AF274003.1 | - | 7 | 1728 | |||
BC001659.2 | - | 8 | 1917 | 2959954 | ||
BC001838.2 | - | 8 | 1917 | 2959954 | ||
BC108316.1 | - | 8 | 2038 | 6151864 |