Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMPDL3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006714.4 | NP_006705.1 | 2 | 477 | ||
XM_005266797.4 | XP_005266854.1 | 2 | 184 | |||
XM_005266798.2 | XP_005266855.1 | 2 | 477 | |||
XM_005266799.2 | XP_005266856.1 | 2 | 477 | |||
XM_017010203.1 | XP_016865692.1 | 2 | 477 | |||
XM_017010204.1 | XP_016865693.1 | 2 | 176 | |||
GenBank mRNA | AK000184.1 | - | 2 | 250 | ||
BC018999.2 | - | 2 | 247 | 3138813 | ||
DA510600.1 | - | 2 | 477 |