Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006717.2 | NP_006708.2 | 5 | 795 | ||
GenBank mRNA | AF087864.1 | - | 5 | 675 | ||
AF106682.1 | - | 5 | 676 | |||
AF317228.2 | - | 4 | 640 | |||
AK092017.1 | - | 8 | 1137 | |||
AK289691.1 | - | 5 | 980 | |||
AK290009.1 | - | 5 | 663 | |||
AK315854.1 | - | 6 | 1143 | |||
AL136719.1 | - | 5 | 758 | |||
BC013571.2 | - | 5 | 779 | |||
BC114515.1 | - | 4 | 650 | 40031486 | ||
BC114565.1 | - | 4 | 650 | 40031487 | ||
BT007314.1 | - | 4 | 480 | |||
CR533484.1 | - | 4 | 480 |
Gene Symbol: BTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BI758136.1 | - | 649 |