Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286474.1 | NP_001273403.1 | 19, 20 | 719 | ||
NM_001286475.1 | NP_001273404.1 | 18, 19 | 698 | |||
NM_006781.4 | NP_006772.3 | 18, 19 | 767 | |||
XM_017010182.1 | XP_016865671.1 | 8, 9 | 189 | |||
XM_017010183.1 | XP_016865672.1 | 8, 9 | 728 | |||
XM_017010184.1 | XP_016865673.1 | 7, 8 | 707 | |||
XM_017010185.1 | XP_016865674.1 | 19, 20 | 792 | |||
GenBank mRNA | AK302176.1 | - | 18, 19 | 658 | ||
AK310230.1 | - | 18, 19 | 687 | |||
BC034774.1 | - | 18, 19 | 734 | 5169157 | ||
U60665.1 | - | 20, 21 | 767 |