Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006785.3 | NP_006776.1 | 5, 6 | 1085 | ||
NM_173844.2 | NP_776216.1 | 5, 6 | 1085 | |||
XM_011525794.1 | XP_011524096.1 | 5, 6 | 1051 | |||
GenBank mRNA | AB026118.1 | - | 5, 6 | 892 | ||
AF130356.2 | - | 5, 6 | 991 | |||
AF316597.1 | - | 5, 6 | 1085 | |||
AK291386.1 | - | 5, 6 | 1051 | |||
BC030143.2 | - | 5, 6 | 939 | 4765195 |