Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320830.1 | NP_001307759.1 | 15 | 1824 | ||
NM_006802.3 | NP_006793.1 | 17 | 1983 | |||
GenBank mRNA | AK128438.1 | - | 17 | 2792 | ||
AK292116.1 | - | 17 | 1894 | |||
BM905122.1 | - | 4 | 671 | |||
U08815.1 | - | 17 | 1846 |