Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034025.1 | NP_001029197.1 | 2 | 764 | ||
NM_006817.3 | NP_006808.1 | 3 | 903 | |||
XM_017018720.1 | XP_016874209.1 | 2 | 772 | |||
GenBank mRNA | BC101493.1 | - | 3 | 843 | 8068999 | |
BC101495.1 | - | 3 | 843 | 8069001 | ||
BE794579.1 | - | 2 | 559 | |||
FJ224317.1 | - | 3 | 796 | |||
X94910.1 | - | 3 | 796 |