Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI44L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006820.3 | NP_006811.2 | 3 | 694 | ||
XM_005270391.3 | XP_005270448.1 | 2 | 201 | |||
XM_005270392.4 | XP_005270449.1 | 2 | 177 | |||
XM_006710304.3 | XP_006710367.1 | 3 | 204 | |||
XM_011540539.2 | XP_011538841.1 | 3 | 532 | |||
XM_017000120.1 | XP_016855609.1 | 3 | 202 | |||
GenBank mRNA | AB000115.1 | - | 3 | 618 | ||
AK223087.1 | - | 3 | 628 | |||
AK225442.1 | - | 3 | 595 | |||
AK293385.1 | - | 3 | 293 | |||
AK316232.1 | - | 2 | 143 | |||
AL832618.2 | - | 1 | 3297 | |||
BC015932.2 | - | 3 | 600 | 3909331 | ||
DC390321.1 | - | 2 | 206 |