Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBNA1BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159936.1 | NP_001153408.1 | 6, 7 | 859 | ||
NM_006824.2 | NP_006815.2 | 5, 6 | 742 | |||
GenBank mRNA | AB451256.1 | - | 5, 6 | 545 | ||
AB451381.1 | - | 5, 6 | 545 | |||
AK223455.1 | - | 5, 6 | 725 | |||
AK299476.1 | - | 3, 4 | 491 | |||
AK307440.1 | - | 5, 6 | 688 | |||
BC009175.2 | - | 5, 6 | 683 | 2966681 | ||
BC073924.1 | - | 11, 12 | 1094 | |||
BT006920.1 | - | 5, 6 | 545 | |||
CR456975.1 | - | 5, 6 | 545 | |||
U86602.1 | - | 5, 6 | 687 |