Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FERMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134999.1 | NP_001128471.1 | 16 | 2415 | ||
NM_006832.2 | NP_006823.1 | 15 | 2394 | |||
XM_005267285.2 | XP_005267342.1 | 16 | 2522 | |||
XM_006720008.2 | XP_006720071.1 | 18 | 2567 | |||
XM_006720009.2 | XP_006720072.1 | 17 | 2543 | |||
XM_006720010.2 | XP_006720073.1 | 17 | 2534 | |||
GenBank mRNA | AF443279.1 | - | 15 | 2289 | ||
AK126426.1 | - | 3 | 1444 | |||
AK291738.1 | - | 15 | 2347 | |||
CR749486.1 | - | 2 | 5077 | |||
Z24725.1 | - | 15 | 2330 |