Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100169.1 | NP_001093639.1 | 3 | 629 | ||
NM_001100170.1 | NP_001093640.1 | 3 | 629 | |||
NM_006839.2 | NP_006830.2 | 3 | 629 | |||
XM_005264110.3 | XP_005264167.1 | 3 | 301 | |||
XM_005264113.1 | XP_005264170.1 | 3 | 297 | |||
XM_005264114.1 | XP_005264171.1 | 3 | 301 | |||
XM_011532497.2 | XP_011530799.1 | 3 | 301 | |||
XM_017003194.1 | XP_016858683.1 | 3 | 297 | |||
XM_017003195.1 | XP_016858684.1 | 3 | 297 | |||
GenBank mRNA | AK225780.1 | - | 3 | 296 | ||
AK296676.1 | - | 3 | 271 | |||
AK299014.1 | - | 3 | 292 | |||
AK299596.1 | - | 1 | 39 | |||
AK300017.1 | - | 3 | 269 | |||
AK304197.1 | - | 3 | 306 | |||
AK309004.1 | - | 3 | 292 | |||
AK312251.1 | - | 3 | 279 | |||
AY232292.1 | - | 3 | 262 | |||
AY921637.1 | - | 3 | 277 | |||
BC002412.2 | - | 3 | 270 | 2964676 | ||
BC005983.1 | - | 3 | 321 | |||
BC107419.1 | - | 3 | 325 | |||
BX537369.1 | - | 3 | 332 | |||
CR456793.1 | - | 3 | 241 | |||
D21091.1 | - | 3 | 326 | |||
D21092.1 | - | 1 | 131 | |||
D21094.1 | - | 3 | 326 | |||
L42572.1 | - | 3 | 333 |