Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100169.1 | NP_001093639.1 | 9 | 1377 | ||
NM_001100170.1 | NP_001093640.1 | 9 | 1347 | |||
NM_006839.2 | NP_006830.2 | 9 | 1380 | |||
XM_005264110.3 | XP_005264167.1 | 9 | 1016 | |||
XM_005264113.1 | XP_005264170.1 | 8 | 952 | |||
XM_005264114.1 | XP_005264171.1 | 8 | 923 | |||
XM_011532497.2 | XP_011530799.1 | 9 | 1040 | |||
XM_017003194.1 | XP_016858683.1 | 9 | 1048 | |||
XM_017003195.1 | XP_016858684.1 | 9 | 1072 | |||
GenBank mRNA | AF148646.1 | - | 5 | 563 | ||
AK225780.1 | - | 9 | 1014 | |||
AK296676.1 | - | 9 | 986 | |||
AK299014.1 | - | 8 | 902 | |||
AK299596.1 | - | 7 | 757 | |||
AK300017.1 | - | 7 | 786 | |||
AK304197.1 | - | 9 | 1021 | |||
AK309004.1 | - | 9 | 1079 | |||
AK312251.1 | - | 9 | 1030 | |||
AY232292.1 | - | 9 | 1010 | |||
AY921637.1 | - | 9 | 1028 | |||
BC002412.2 | - | 9 | 1021 | 2964676 | ||
BX537369.1 | - | 9 | 1050 | |||
CR456793.1 | - | 9 | 992 | |||
CR749590.1 | - | 4 | 424 | |||
D21092.1 | - | 7 | 882 | |||
D21094.1 | - | 9 | 1077 | |||
L42572.1 | - | 9 | 1084 |