Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100169.1 | NP_001093639.1 | 14 | 1983 | ||
NM_001100170.1 | NP_001093640.1 | 14 | 1953 | |||
NM_006839.2 | NP_006830.2 | 14 | 1986 | |||
XM_005264110.3 | XP_005264167.1 | 14 | 1622 | |||
XM_005264113.1 | XP_005264170.1 | 13 | 1558 | |||
XM_005264114.1 | XP_005264171.1 | 13 | 1529 | |||
XM_011532497.2 | XP_011530799.1 | 14 | 1646 | |||
XM_017003194.1 | XP_016858683.1 | 14 | 1654 | |||
XM_017003195.1 | XP_016858684.1 | 14 | 1678 | |||
GenBank mRNA | AF148646.1 | - | 9 | 1154 | ||
AK225780.1 | - | 14 | 1620 | |||
AK296676.1 | - | 14 | 1592 | |||
AK299014.1 | - | 13 | 1508 | |||
AK299596.1 | - | 12 | 1363 | |||
AK300017.1 | - | 12 | 1329 | |||
AK312251.1 | - | 14 | 1636 | |||
AY232292.1 | - | 14 | 1616 | |||
AY921637.1 | - | 14 | 1634 | |||
BC002412.2 | - | 14 | 1627 | 2964676 | ||
BC027458.1 | - | 3 | 339 | |||
BX537369.1 | - | 14 | 1657 | |||
CR456793.1 | - | 14 | 1598 | |||
CR749590.1 | - | 9 | 1030 | |||
D21092.1 | - | 12 | 1488 | |||
D21093.1 | - | 3 | 250 | |||
D21094.1 | - | 14 | 1683 | |||
L42572.1 | - | 14 | 1690 |