Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290049.1 | NP_001276978.1 | 6, 7 | 714 | ||
NM_003715.3 | NP_003706.2 | 5, 6 | 702 | |||
XM_006714396.3 | XP_006714459.1 | 5, 6 | 697 | |||
XM_006714397.3 | XP_006714460.1 | 6, 7 | 714 | |||
GenBank mRNA | AK314289.1 | - | 5, 6 | 592 | ||
AL832010.2 | - | 6, 7 | 637 | |||
BC006398.2 | - | 5, 6 | 627 | |||
BC032654.1 | - | 5, 6 | 557 | 5533256 | ||
D86326.1 | - | 5, 6 | 579 |