Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TLK2 SIRNA Assays ›
Gene Symbol:
TLK2
Gene Name:
tousled like kinase 2
Gene Aliases:
HsHPK, PKU-ALPHA
Chromosome Location:
Chr.17: 62477481 - 62615481 on Build GRCh38
UniGene ID:
Hs.445078
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136136
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TLK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284333.1
NP_001271262.1
23
3124
NM_001284363.1
NP_001271292.1
20
2719
NM_006852.3
NP_006843.2
22
3061
XM_011524215.2
XP_011522517.1
21
2768
XM_011524216.2
XP_011522518.1
23
2888
XM_011524217.2
XP_011522519.1
23
3075
XM_011524218.2
XP_011522520.1
20
2702
XM_011524220.2
XP_011522522.1
20
2672
XM_011524221.2
XP_011522523.1
22
2786
XM_011524222.2
XP_011522524.1
23
3267
XM_011524223.2
XP_011522525.1
21
2929
XM_011524224.2
XP_011522526.1
21
2987
XM_011524225.2
XP_011522527.1
21
2935
XM_011524226.2
XP_011522528.1
23
2981
XM_011524227.2
XP_011522529.1
22
3035
XM_011524228.2
XP_011522530.1
23
3135
XM_011524229.2
XP_011522531.1
24
3062
XM_017024044.1
XP_016879533.1
22
2895
XM_017024045.1
XP_016879534.1
21
3003
XM_017024046.1
XP_016879535.1
22
2825
XM_017024047.1
XP_016879536.1
22
2822
XM_017024048.1
XP_016879537.1
22
3283
XM_017024049.1
XP_016879538.1
22
3236
XM_017024050.1
XP_016879539.1
20
2768
XM_017024051.1
XP_016879540.1
21
3217
XM_017024052.1
XP_016879541.1
21
3168
XM_017024053.1
XP_016879542.1
21
2840
XM_017024054.1
XP_016879543.1
20
2866
XM_017024055.1
XP_016879544.1
19
2770
XM_017024056.1
XP_016879545.1
20
2921
XM_017024057.1
XP_016879546.1
20
2735
XM_017024058.1
XP_016879547.1
22
3013
XM_017024059.1
XP_016879548.1
22
3003
XM_017024060.1
XP_016879549.1
21
2920
GenBank mRNA
AB004884.1
-
20
2697
AF162667.1
-
22
2933
AX775745.1
-
22
2933
BC044925.1
-
23
3031
BX647225.1
-
1
1157
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of chromatin assembly or disassembly
protein phosphorylation
cellular response to DNA damage stimulus
cell cycle
chromosome segregation
negative regulation of autophagy
covalent chromatin modification
peptidyl-serine phosphorylation
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
intracellular signal transduction
cellular response to gamma radiation
protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-g5d4h:80/100.66.76.71:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline