Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007006.2 | NP_008937.1 | 4 | 606 | ||
GenBank mRNA | AJ001810.1 | - | 4 | 459 | ||
AK312721.1 | - | 4 | 609 | |||
AL535307.3 | - | 4 | 606 | |||
BC001403.2 | - | 4 | 567 | 3139379 | ||
BX537360.1 | - | 4 | 610 | |||
CR456919.1 | - | 4 | 459 |