Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317099.1 | NP_001304028.1 | 4 | 1093 | ||
NM_001317100.1 | NP_001304029.1 | 4 | 1287 | |||
NM_001317101.1 | NP_001304030.1 | 3 | 1174 | |||
NM_001317102.1 | NP_001304031.1 | 3 | 848 | |||
NM_001317103.1 | NP_001304032.1 | 2 | 637 | |||
NM_007034.4 | NP_008965.2 | 3 | 1206 | |||
GenBank mRNA | AB209922.1 | - | 3 | 1084 | ||
AK297985.1 | - | 3 | 756 | |||
AK313205.1 | - | 3 | 1092 | |||
BC034721.1 | - | 3 | 1082 | 4340658 | ||
BI910092.1 | - | 2 | 529 | |||
U40992.2 | - | 3 | 1099 |