Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317099.1 | NP_001304028.1 | 4 | 1330 | ||
NM_001317100.1 | NP_001304029.1 | 4 | 1524 | |||
NM_001317101.1 | NP_001304030.1 | 3 | 1411 | |||
NM_001317102.1 | NP_001304031.1 | 3 | 1085 | |||
NM_001317103.1 | NP_001304032.1 | 2 | 874 | |||
NM_007034.4 | NP_008965.2 | 3 | 1443 | |||
GenBank mRNA | AB209922.1 | - | 3 | 1321 | ||
BC034721.1 | - | 3 | 1319 | 4340658 |