Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003725.3 | NP_003716.2 | 3 | 608 | ||
XM_005269207.1 | XP_005269264.1 | 4 | 757 | |||
XM_005269208.1 | XP_005269265.1 | 4 | 761 | |||
XM_005269209.1 | XP_005269266.1 | 4 | 748 | |||
XM_006719672.1 | XP_006719735.1 | 4 | 756 | |||
XM_011538925.1 | XP_011537227.1 | 4 | 751 | |||
XM_011538926.1 | XP_011537228.1 | 4 | 741 | |||
XM_011538927.1 | XP_011537229.1 | 4 | 732 | |||
GenBank mRNA | AF016509.1 | - | 2 | 392 | ||
AF223225.1 | - | 3 | 479 | |||
AK313423.1 | - | 3 | 518 | |||
BC020710.1 | - | 3 | 502 | 4700726 | ||
U89281.1 | - | 3 | 497 |