Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KERA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007035.3 | NP_008966.1 | 2 | 915 | ||
GenBank mRNA | AF063301.1 | - | 1 | 304 | ||
AF205403.1 | - | 2 | 544 | |||
AK314990.1 | - | 2 | 477 | |||
BC032667.2 | - | 1 | 305 | 5541591 | ||
BM722433.1 | - | 2 | 448 | |||
CV571245.1 | - | 2 | 462 |