Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102426.1 | NP_001095896.1 | 14 | 2479 | ||
XM_005263862.3 | XP_005263919.1 | 14 | 2768 | |||
XM_005263863.4 | XP_005263920.1 | 14 | 2555 | |||
XM_005263864.3 | XP_005263921.1 | 14 | 2720 | |||
XM_011510507.1 | XP_011508809.1 | 16 | 2610 | |||
XM_017003198.1 | XP_016858687.1 | 13 | 2675 | |||
XM_017003199.1 | XP_016858688.1 | 13 | 2628 | |||
GenBank mRNA | AB017114.1 | - | 10 | 1607 | ||
AB449889.1 | - | 14 | 2348 | |||
AK292826.1 | - | 12 | 2112 | |||
BQ673509.1 | - | 5 | 697 |