Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007065.3 | NP_008996.1 | 2 | 469 | ||
XM_011527652.2 | XP_011525954.1 | 2 | 436 | |||
GenBank mRNA | BC000083.2 | - | 2 | 393 | 3505011 | |
BC008793.2 | - | 2 | 393 | 3619769 | ||
BE796008.1 | - | 2 | 473 | |||
BT006796.1 | - | 2 | 353 | |||
CR542249.1 | - | 2 | 353 | |||
U43077.1 | - | 2 | 361 | |||
U63131.1 | - | 2 | 401 |