Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HHLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282556.1 | NP_001269485.1 | 6 | 945 | ||
NM_001282557.1 | NP_001269486.1 | 6 | 876 | |||
NM_001282558.1 | NP_001269487.1 | 7 | 1023 | |||
NM_001282559.1 | NP_001269488.1 | 7 | 761 | |||
NM_007072.3 | NP_009003.1 | 6 | 1136 | |||
XM_005247080.3 | XP_005247137.1 | 7 | 1006 | |||
XM_011512362.1 | XP_011510664.1 | 7 | 1006 | |||
XM_011512363.1 | XP_011510665.1 | 6 | 936 | |||
XM_011512364.1 | XP_011510666.1 | 6 | 887 | |||
XM_011512367.2 | XP_011510669.1 | 7 | 1006 | |||
XM_017005642.1 | XP_016861131.1 | 6 | 936 | |||
GenBank mRNA | AF126162.1 | - | 6 | 876 | ||
AK000692.1 | - | 5 | 871 | |||
AK027132.1 | - | 3 | 429 | |||
AK296644.1 | - | 7 | 704 | |||
AK315109.1 | - | 7 | 968 | |||
BC035971.1 | - | 6 | 1136 | 4606764 |