Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193333.2 | NP_001180262.1 | 9 | 1312 | ||
NM_007074.3 | NP_009005.1 | 8 | 1194 | |||
XM_011545714.2 | XP_011544016.1 | 8 | 1007 | |||
XM_017022885.1 | XP_016878374.1 | 8 | 1182 | |||
XM_017022886.1 | XP_016878375.1 | 8 | 1004 | |||
GenBank mRNA | AB209221.1 | - | 3 | 3474 | ||
AF495470.1 | - | 8 | 999 | |||
AK096332.1 | - | 7 | 2189 | |||
AK097964.1 | - | 2 | 1118 | |||
AK123401.1 | - | 3 | 1773 | |||
AK314714.1 | - | 9 | 1113 | |||
BC110374.1 | - | 8 | 1011 | 5733853 | ||
BC126385.1 | - | 8 | 898 | 8992101 | ||
BC126387.1 | - | 8 | 898 | 8992103 | ||
D44497.1 | - | 8 | 977 | |||
U34690.1 | - | 8 | 969 | |||
X89109.1 | - | 8 | 970 |