Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007085.4 | NP_009016.1 | 11 | 1094 | ||
GenBank mRNA | AF288537.1 | - | 5 | 374 | ||
AK289388.1 | - | 11 | 1015 | |||
AK291138.1 | - | 11 | 1016 | |||
AK300356.1 | - | 10 | 909 | |||
AK300436.1 | - | 11 | 945 | |||
BC000055.2 | - | 11 | 1001 | 3505833 | ||
BX647421.1 | - | 11 | 1019 | |||
U06863.1 | - | 11 | 1010 |