Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007085.4 | NP_009016.1 | 11 | 2276 | ||
GenBank mRNA | AK025860.1 | - | 1 | 145 | ||
BC000055.2 | - | 11 | 2184 | 3505833 | ||
BC017413.1 | - | 1 | 60 | |||
BX647421.1 | - | 11 | 2201 |