Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007178.3 | NP_009109.3 | 6 | 972 | ||
XM_005253294.1 | XP_005253351.1 | 5 | 834 | |||
GenBank mRNA | AB024327.1 | - | 6 | 918 | ||
AF161496.1 | - | 6 | 919 | |||
AJ010025.1 | - | 6 | 765 | |||
AK297942.1 | - | 7 | 971 | |||
AK312295.1 | - | 6 | 913 | |||
AK315891.1 | - | 5 | 799 | |||
AL136691.1 | - | 6 | 914 | |||
AY049776.1 | - | 6 | 665 | |||
BC000162.2 | - | 6 | 763 | 2900954 | ||
BC062306.1 | - | 6 | 927 | 3349549 | ||
BT020044.1 | - | 6 | 618 | |||
BT020045.1 | - | 6 | 618 |