Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172648.3 | NP_001166119.1 | 11 | 1776 | ||
NM_001172649.3 | NP_001166120.1 | 10 | 1873 | |||
NM_001291106.1 | NP_001278035.1 | 11 | 2020 | |||
NM_001291107.1 | NP_001278036.1 | 11 | 2020 | |||
NM_007185.6 | NP_009116.3 | 11 | 2023 | |||
XM_006711139.2 | XP_006711202.1 | 12 | 1251 | |||
XM_006711141.2 | XP_006711204.1 | 12 | 1248 | |||
XM_011509111.2 | XP_011507413.1 | 10 | 1718 | |||
XM_011509112.2 | XP_011507414.1 | 10 | 1114 | |||
XM_011509114.2 | XP_011507416.1 | 10 | 1082 | |||
XM_017000145.1 | XP_016855634.1 | 10 | 1089 | |||
XM_017000146.1 | XP_016855635.1 | 10 | 1086 | |||
GenBank mRNA | AB209264.1 | - | 11 | 1776 | ||
AF284423.1 | - | 2 | 250 | |||
AF329264.1 | - | 11 | 1214 | |||
AK298852.1 | - | 5 | 646 | |||
AY165003.1 | - | 11 | 2013 | |||
BC041088.1 | - | 10 | 1105 | |||
BC052491.1 | - | 11 | 1994 | 4820737 | ||
BC068008.1 | - | 10 | 1050 | |||
BC104758.1 | - | 11 | 1286 | 8143761 | ||
BC143226.1 | - | 11 | 1283 | 9051727 | ||
U80746.1 | - | 8 | 902 | |||
U80759.1 | - | 8 | 909 |