Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318219.1 | NP_001305148.1 | 9, 10 | 1272 | ||
NM_007186.5 | NP_009117.2 | 11, 12 | 1693 | |||
XM_005260262.4 | XP_005260319.1 | 11, 12 | 1446 | |||
XM_005260263.4 | XP_005260320.1 | 11, 12 | 1446 | |||
XM_005260264.4 | XP_005260321.1 | 11, 12 | 1446 | |||
XM_006723690.3 | XP_006723753.1 | 11, 12 | 1443 | |||
XM_006723691.1 | XP_006723754.1 | 11, 12 | 1659 | |||
XM_006723692.3 | XP_006723755.1 | 10, 11 | 1369 | |||
XM_006723693.3 | XP_006723756.1 | 10, 11 | 1353 | |||
XM_006723694.3 | XP_006723757.1 | 11, 12 | 1446 | |||
XM_011528517.2 | XP_011526819.1 | 11, 12 | 1446 | |||
XM_011528518.2 | XP_011526820.1 | 11, 12 | 1450 | |||
XM_011528519.2 | XP_011526821.1 | 4, 5 | 559 | |||
XM_017027617.1 | XP_016883106.1 | 11, 12 | 1446 | |||
XM_017027618.1 | XP_016883107.1 | 3, 4 | 546 | |||
GenBank mRNA | AF022655.1 | - | 9, 10 | 1267 | ||
AF049105.1 | - | 12, 13 | 1717 | |||
AK297572.1 | - | 10, 11 | 1427 | |||
BC144706.1 | - | 9, 10 | 1218 |