Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286124.1 | NP_001273053.1 | 13 | 2093 | ||
NM_001286126.1 | NP_001273055.1 | 13 | 2093 | |||
NM_001286128.1 | NP_001273057.1 | 12 | 2027 | |||
NM_018650.4 | NP_061120.3 | 13 | 2093 | |||
XM_005273134.4 | XP_005273191.1 | 13 | 1960 | |||
XM_006711326.3 | XP_006711389.1 | 13 | 1354 | |||
XM_011509561.2 | XP_011507863.1 | 13 | 2089 | |||
XM_011509562.2 | XP_011507864.1 | 13 | 1706 | |||
XM_017001305.1 | XP_016856794.1 | 14 | 1550 | |||
XM_017001306.1 | XP_016856795.1 | 13 | 1590 | |||
XM_017001307.1 | XP_016856796.1 | 14 | 1990 | |||
GenBank mRNA | AB040910.1 | - | 12 | 1984 | ||
AF154845.1 | - | 13 | 1719 | |||
AK027493.1 | - | 9 | 1050 | |||
AK295506.1 | - | 13 | 1591 | |||
AK314614.1 | - | 13 | 1753 | |||
BC113869.1 | - | 13 | 1629 | 40036731 | ||
BC114478.1 | - | 13 | 1629 | 40036730 | ||
KJ534879.1 | - | 13 | 1453 | |||
KJ534880.1 | - | 7 | 853 | |||
KJ534881.1 | - | 13 | 1453 | |||
KJ535089.1 | - | 13 | 1452 |